Skip to Content
MilliporeSigma

BL3H1F1CA489

BLD Pharmatech

[1,1'-Binaphthalene]-2,2'-diamine

Synonym(s):

1,1'-binaphthyl-2,2'-diamine

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C20H16N2
CAS Number:
Molecular Weight:
284.36
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C20H16N2/c21-17-11-9-13-5-1-3-7-15(13)19(17)20-16-8-4-2-6-14(16)10-12-18(20)22/h1-12H,21-22H2

Inchi Key

DDAPSNKEOHDLKB-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

97%

Storage temp.

Keep in dark place, sealed in dry, room temperature

Country of Origin

CN

Physical Form

Form: Crystal - Powder / Colour: White - Pale yellow red - Brown

Melting Point

240-244C



Still not finding the right product?

Explore all of our products under [1,1'-Binaphthalene]-2,2'-diamine


signalword

Warning

pictograms

Exclamation mark


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library