Skip to Content
MilliporeSigma

BL3H1F1CD5CF

BLD Pharmatech

2-Bromo-1-(4-(dimethylamino)phenyl)ethanone

Synonym(s):

2-bromo-1-[4-(dimethylamino)phenyl]ethanone

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C10H12O1N1Br1
CAS Number:
Molecular Weight:
242.12
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Storage temp.

Inert atmosphere, 2-8°C

Inchi Code

1S/C10H12BrNO/c1-12(2)9-5-3-8(4-6-9)10(13)7-11/h3-6H,7H2,1-2H3

Inchi Key

YWNKTZFJKMGVRP-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

95%

Country of Origin

CN

Physical Form

Solid



Still not finding the right product?


signalword

Warning

pictograms

Exclamation mark


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library