Skip to Content
MilliporeSigma

BL3H97A466B5

BLD Pharmatech

4'-Bromospiro[adamantane-2,9'-fluorene]

Synonym(s):

(1R,3S,5r,7r)-4'-bromospiro[adamantane-2,9'-fluorene]

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C22H21Br1
CAS Number:
Molecular Weight:
365.31
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C22H21Br/c23-20-7-3-6-19-21(20)17-4-1-2-5-18(17)22(19)15-9-13-8-14(11-15)12-16(22)10-13/h1-7,13-16H,8-12H2

Inchi Key

QGQSZWNWWFSYMK-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

97%

Storage temp.

Sealed in dry, room temperature

Country of Origin

CN

Physical Form

Solid



Still not finding the right product?




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library