Skip to Content
MilliporeSigma

BL3H97A46F0C

BLD Pharmatech

Dodecan-1-amine

Synonym(s):

1-dodecanamine

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C12H27N1
CAS Number:
Molecular Weight:
185.35
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C12H27N/c1-2-3-4-5-6-7-8-9-10-11-12-13/h2-13H2,1H3

Inchi Key

JRBPAEWTRLWTQC-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

98%

Storage temp.

Keep in dark place, inert atmosphere, room temperature

Country of Origin

CN

Physical Form

Colorless to White to Yellow Solid or Semi-solid or liquid



Still not finding the right product?

Explore all of our products under Dodecan-1-amine




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library