Skip to Content
MilliporeSigma

BL3H97A488D1

BLD Pharmatech

Nicotinoyl chloride hydrochloride

Synonym(s):

nicotinoyl chloride hydrochloride

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C6H5O1N1Cl2
CAS Number:
Molecular Weight:
178.02
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C6H4ClNO.ClH/c7-6(9)5-2-1-3-8-4-5;/h1-4H;1H

Inchi Key

MSYBLBLAMDYKKZ-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

95%

Storage temp.

Inert atmosphere, 2-8C

Country of Origin

CN

Physical Form

White to off-white powder or crystals

Melting Point

151-157C



Still not finding the right product?

Explore all of our products under Nicotinoyl chloride hydrochloride


hstatements

signalword

Danger

pictograms

Corrosion


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library