Skip to Content
MilliporeSigma

BL3H9A625576

BLD Pharmatech

(2S,3S)-2-((S)-1-((2S,3S)-2-Amino-3-methylpentanoyl)pyrrolidine-2-carboxamido)-3-methylpentanoic acid

Synonym(s):

(2S,3S)-2-[({(2S)-1-[(2S,3S)-2-amino-3-methylpentanoyl]-2-pyrrolidinyl}carbonyl)amino]-3-methylpentanoic acid

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C17H31O4N3
CAS Number:
Molecular Weight:
341.45
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C17H31N3O4/c1-5-10(3)13(18)16(22)20-9-7-8-12(20)15(21)19-14(17(23)24)11(4)6-2/h10-14H,5-9,18H2,1-4H3,(H,19,21)(H,23,24)/t10-,11-,12-,13-,14-/m0/s1

Inchi Key

JNTMAZFVYNDPLB-PEDHHIEDSA-N

Shipping Temperature

Normal

Purity

99%

Storage temp.

Sealed in dry, store in freezer, under -20C

Country of Origin

CN

Physical Form

Solid





Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library