Skip to Content
MilliporeSigma

CLR-VDHCH-CR-CC-8540

Blue Ridge Ergonomics

Blue Ridge Ergonomics® Class 10 Cleanroom Black Vinyl Lab Chair Standard Back

Desk Height

Shipped & Sold By Blue Ridge Ergonomics

A Trusted Marketplace Seller

Synonym(s):

cleanroom chair, lab chair, class 10, vinyl chair

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

Marketplace Support
Need help or have questions about this product?
Contact Seller


weight

31 lb

H × W × D

34 in x 25 in x 25 in

manufacturer/tradename

E Com Inc.

General description

Our Class 10 vinyl desk height clean room chair in chrome is adjustable from 19-24 inches from the top of the seat to the floor and features ergonomic adjustments including seat height and locking back tilt and cast-aluminum base with chromehooded, dual-wheel, self-braking, non-marring casters suitable for hard floors in Black Trailblazer™ Vinyl.

  • Laboratory-Grade Vinyl.
  • Meets Class 10 Cleanroom Standards (ISO4).
  • Features Stainless Steel Hardware.
  • Easy to Clean.
  • COLOR Black.
  • WEIGHT CAPACITY 300lbs.
  • WARRANTY 15-Years on Parts; Lifetime for Pneumatic Cylinders.
  • Made in the USA


Still not finding the right product?




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library