Skip to Content
MilliporeSigma

HPDHCH-RG-T0-A0-RC-GRY

Blue Ridge Ergonomics

Blue Ridge Ergonomics® Desk Height Lab Chair Black Base with Integrated Handle and Casters

Gray Polyurethane

Shipped & Sold By Blue Ridge Ergonomics

A Trusted Marketplace Seller

Synonym(s):

lab chair, polyurethane chair, seating

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

Marketplace Support
Need help or have questions about this product?
Contact Seller


weight

31 lb

H × W × D

33 in x 26 in x 26 in

manufacturer/tradename

E Com Inc.

General description

Our gray polyurethane desk height chair is adjustable from 17.25-22.5 inches from the top of the seat to the floor and features ergonomic adjustments including seat height and locking back tilt, built in handle for ease of movement, and black, nylon-reinforced fiberglass base with dual-wheel, self-braking, non-marring casters suitable for hard floors in chemical, soil, and puncture resistant polyurethane.

  • Chemical Resistant Polyurethane.
  • Enhanced Durability.
  • COLOR Gray.
  • WEIGHT CAPACITY 300lbs.
  • WARRANTY 15-Years on Parts; Lifetime for Pneumatic Cylinders.
  • Made in the USA





Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library