Skip to Content
MilliporeSigma

MOD-MDHCH-RG-C2-A0-RC

Blue Ridge Ergonomics

Blue Ridge Ergonomics®Oxford Mesh Back Chair

Desk Height

Shipped & Sold By Blue Ridge Ergonomics

A Trusted Marketplace Seller

Synonym(s):

office chair, office furniture, mesh back chair

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

Marketplace Support
Need help or have questions about this product?
Contact Seller


weight

60 lb

H × W × D

37 in x 27 in x 27 in

manufacturer/tradename

E Com Inc.

General description

Our Oxford mesh back chair is adjustable from 19-23 inches from the top of the seat to the floor and features adjustable lumbar support, asynchronous knee-bend mechanism, and composite base with dual-wheel casters suitable for carpeted f loors..

  • NOT SUITABLE FOR LAB ENVIRONMENT.
  • Adjustable Lumbar Support
  • COLOR Black.
  • WEIGHT CAPACITY 300lbs.
  • WARRANTY 5 year parts, lifetime on pneumatic lift, 5 year on upholstery
  • Made in the USA




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library