Skip to Content
MilliporeSigma

PTHBST-CR-CF-RG-BLK

Blue Ridge Ergonomics

Blue Ridge Ergonomics® Black Polyurethane Tractor Stool

High Bench Height

Shipped & Sold By Blue Ridge Ergonomics

A Trusted Marketplace Seller

Synonym(s):

lab stool, polyurethane stool, seating

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

Marketplace Support
Need help or have questions about this product?
Contact Seller


weight

29 lb

H × W × D

23 in x 25 in x 25 in

manufacturer/tradename

E Com Inc.

General description

Our black polyurethane high bench height tractor stool in chrome is adjustable from 23-33.5 inches from the top of the seat to the floor and features ergonomic adjustments such as seat height and locking seat tilt, chrome adjustable foot ring, and cast-aluminum base with stationary glides suitable for hard floors in chemical in chemical, soil, and puncture resistant polyurethane.

  • Chemical Resisant Polyurethane.
  • Enhanced Durability.
  • COLOR Black.
  • FOOTRING Chrome.
  • WEIGHT CAPACITY 300lbs.
  • WARRANTY 15-Years on Parts; Lifetime for Pneumatic Cylinders.
  • Made in the USA


Still not finding the right product?




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library