Skip to Content
MilliporeSigma

VDHSO-CR-CC-8540

Blue Ridge Ergonomics

Blue Ridge Ergonomics® Black Vinyl Stool

Desk Height

Shipped & Sold By Blue Ridge Ergonomics

A Trusted Marketplace Seller

Synonym(s):

lab stool, vinyl stool, seating

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

Marketplace Support
Need help or have questions about this product?
Contact Seller


weight

25 lb

H × W × D

18 in x 25 in x 25 in

manufacturer/tradename

E Com Inc.

General description

Our vinyl desk height stool is adjustable from 18.25-23.25 inches from the top of the seat to the floor and features ergonomic adjustments, including 360- degree, all-around control, and aluminum base with chrome-hooded, dual-wheel, self-braking, non-marring casters suitable for hard floors in Black Trailblazer™ Vinyl.

  • Laboratory-Grade Vinyl.
  • Easy to Clean.
  • COLOR Black.
  • WEIGHT CAPACITY 300lbs.
  • WARRANTY 15-Years on Parts; Lifetime for Pneumatic Cylinders.
  • Made in the USA


Still not finding the right product?




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library