Skip to Content
MilliporeSigma

VMBCH-MB-CR-T1-A1-CF-CC-8823

Blue Ridge Ergonomics

Blue Ridge Ergonomics® Carbon Vinyl Lab Chair Chrome Base with Large Back, Seat Tilt, Arms, Chrome Foot Ring, and Casters

Medium Bench Height

Shipped & Sold By Blue Ridge Ergonomics

A Trusted Marketplace Seller

Synonym(s):

lab chair, vinyl chair, seating

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

Marketplace Support
Need help or have questions about this product?
Contact Seller


weight

45 lb

H × W × D

42 in x 26 in x 26 in

manufacturer/tradename

E Com Inc.

General description

Our vinyl medium bench height chair with large back in chrome is adjustable from 21.5-29 inches from the top of the seat to the floor and features ergonomic adjustments including seat height and locking back tilt, locking seat tilt, 6-way adjustable arm rests, chrome adjustable foot ring, and cast-aluminum base with chrome-hooded, dual-wheel, self-braking, non-marring casters suitable for hard floors in Carbon Supernova™ Vinyl.

  • Laboratory-Grade Vinyl.
  • Easy to Clean.
  • COLOR Carbon.
  • Locking Seat Tilt.
  • 6-Way Adjustable Arms.
  • FOOTRING Chrome.
  • WEIGHT CAPACITY 300lbs.
  • WARRANTY 15-Years on Parts; Lifetime for Pneumatic Cylinders; 5-Years on Upholstery.
  • Made in the USA




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library