MilliporeSigma
CAPL-222-3902-085
Caplugs® Evergreen
Caplugs® Evergreen CryoSure® Cryogenic Vials with Caps
1.5 mL CryoSure® Cryogenic Freestanding Vials with Natural Screw Caps, Sterile, Polypropylene Co-Polymer, Natural, 500/Bag
Shipped & Sold By Caplugs, Inc.
A Trusted Marketplace Seller
Synonym(s):
Cryogenic Vials, Small Sample Vials
Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
Marketplace Support
Need help or have questions about this product?
Contact Seller
material
polypropylene copolymer
O.D.
12.25 mm
volume
1.5 mL, 1500 μL
manufacturer/tradename
Caplugs® Evergreen
General description
Caplugs® Evergreen CryoSure® Cryogenic Vials are one of the two styles of small sample vials offered by our catalog, ideal for long-term storage of biological samples at ultra-low temperatures. Made from durable polypropylene co-polymer, these vials are sterile, leak-proof, and designed to withstand temperatures as low as vapor phase* liquid nitrogen (-320°F/-195°C). Perfect for use in biobanks, research labs, and clinical settings, storing blood/serum samples, cell cultures, sperm, and other biological specimens. What makes this style of vial different is the uniform diameter and straight vertical inside walls of the vial with no shoulder to allow for a large accessible opening which makes processing and retrieval of samples convenient.
  • 95 kPa compliant
  • Gamma irradiated sterilization
  • Molded-in graduations every 0.5 mL
  • External threads to minimize sample contamination
  • Uniform diameter of all vials
*Caplugs does not recommend the use of CryoSure® Cryogenic Vials for immersion in liquid nitrogen or any other liquefied gases. Immersion may cause potential leakage in vials with the subsequent danger of rapid pressure build-up and/or explosion when returned to room temperature. Please use CryoSure® Cryogenic Vials with caution.
Still not finding the right product?
Choose from one of the most recent versions:
Certificates of Analysis (COA)