MilliporeSigma
CAPL-240-3074-Z30
Caplugs® Evergreen
Caplugs® Evergreen Five-Slide Mailers
Five-Slide Mailers, Non-Sterile, Polypropylene, Black, 100/Bag
Shipped & Sold By Caplugs, Inc.
A Trusted Marketplace Seller
Synonym(s):
Slide Mailers
Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
Marketplace Support
Need help or have questions about this product?
Contact Seller
material
polypropylene
W × L
18 mm x 89 mm
manufacturer/tradename
Caplugs® Evergreen
General description
Caplugs® Evergreen Five-Slide Mailers feature an integral recessed lid and molded-in dividers to limit slide movement during transit to protect against accidental breakage. These mailers are designed with rounded corners to allow easier handling and an eyelet for the optional security ties and/or identification tags. All these aspects allow for more secure and convenient shipping, providing the protection that laboratories require for microscope slides during transport. Ensure your slide samples arrive safely and intact!
  • Lightweight (less than 1/3 oz) to help minimize transport costs
  • Mailers can also function as Coplin jars, similar to our Slide-Fix™ Jars
Still not finding the right product?
Choose from one of the most recent versions:
Certificates of Analysis (COA)