MilliporeSigma
CAPL-240-3800-Y30
Caplugs® Evergreen
Caplugs® Evergreen Slide-Sette Savers™
Slide-Sette Saver™ Microscope Slide & Paraffin Block Storage Containers, Non-Sterile, Polypropylene, Yellow, 100/Bag
Shipped & Sold By Caplugs, Inc.
A Trusted Marketplace Seller
Synonym(s):
Slide Storage
Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
Marketplace Support
Need help or have questions about this product?
Contact Seller
material
polypropylene
W × L
20 mm x 84 mm
manufacturer/tradename
Caplugs® Evergreen
General description
Caplugs® Evergreen Slide-Sette Savers™ are designed for the long-term storage and shipment of multiple pathology samples from the same patient in one singular container. Use our uniquely-designed product with protective lid and living hinges to cocoon your microscope slides and cassette/paraffin blocks inside their respective compartments with a high degree of protection and safety. Multiple samples collected with one container to hold them all, making testing easier!
  • Manufactured under license from the Moffitt Cancer Center
  • Available in multiple colors for potential color-coding of malignant tissues collected, for example, pink for breast cancer, yellow for bladder cancer, etc.
Still not finding the right product?
Choose from one of the most recent versions:
Certificates of Analysis (COA)