MilliporeSigma
CAPL-240-3828-W80
Caplugs® Evergreen
Caplugs® Evergreen Slide-Sette Saver™ Storage Boxes
Slide-Sette Saver™ Storage Boxes, 20 Positions, Non-Sterile, Polypropylene Co-Polymer, White, 3/Pack
Shipped & Sold By Caplugs, Inc.
A Trusted Marketplace Seller
Synonym(s):
Slide Storage
Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
Marketplace Support
Need help or have questions about this product?
Contact Seller
material
polypropylene copolymer
manufacturer/tradename
Caplugs® Evergreen
General description
Caplugs® Evergreen Slide-Sette Saver™ Storage Box is designed to hold multiple of our Slide-Sette Savers™, used for the storage of multiple pathology specimens collected from the same patient. Each box features hinged transparent polypropylene lid with a colored high impact polystyrene bottom with dual latches on every box to provide secured transport and storage.
  • Slide-Sette Saver™ Capacity: 20
  • 2 x 10 configuration
  • Features transparent polypropylene lid with a hinged bottom made of high impact polystyrene
Still not finding the right product?
Choose from one of the most recent versions:
Certificates of Analysis (COA)