MilliporeSigma
CAPL-240-7020-B80
Caplugs® Evergreen
Caplugs® Evergreen Histo-Click™ Specimen Containers with Caps
Histo-Click™ 20 mL Click-Top Specimen Containers with Blue Caps, Non-Sterile, Polypropylene Co-Polymer, Natural, 1000/Carton Unassembled
Shipped & Sold By Caplugs, Inc.
A Trusted Marketplace Seller
Synonym(s):
Specimen Container, Click Top
Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
Marketplace Support
Need help or have questions about this product?
Contact Seller
material
polypropylene copolymer
O.D.
35.5 mm
volume
20 mL, 20000 μL
manufacturer/tradename
Caplugs® Evergreen
General description
Listen for the click! Caplugs® Evergreen Histo-Click™ Specimen Containers are designed for secured storage of biological fluids and pathology specimens in a container that provides extra assurance of a secure, tight seal with it′s audible click seal. These multipurpose specimen containers are ideal for transporting samples, storing tissue specimens, and perfect for storage of pathological and cyto-histo specimens once filled with formalin, ethyl alcohol or other reagents.
  • 95 kPa & thermal shock compliant
  • Molded-in graduations
  • Nestable design for optimal storage
Still not finding the right product?
Choose from one of the most recent versions:
Certificates of Analysis (COA)