MilliporeSigma
CAPL-300-3900-L20
Caplugs® Evergreen
Caplugs® Evergreen CryoSure® Cryogenic Vial Caps
CryoSure® Cryogenic Vial Caps, Non-Sterile, HDPE, Lavender, 200/Bag
Shipped & Sold By Caplugs, Inc.
A Trusted Marketplace Seller
Synonym(s):
Cryogenic Vials, Small Sample Vials
Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
Marketplace Support
Need help or have questions about this product?
Contact Seller
material
high-density polyethylene
O.D.
12.25 mm
manufacturer/tradename
Caplugs® Evergreen
General description
Caplugs® Evergreen CryoSure® Caps are designed to fit all sizes of our CryoSure® cryogenic vials. Our caps molded-in sealing ring provides a leakproof closure without the need for a silicone gasket, making them ideal for cryogenic storage.
  • Available in a variety of colors
  • Vials with caps are 95 kPa compliant
Still not finding the right product?
Choose from one of the most recent versions:
Certificates of Analysis (COA)