MilliporeSigma
CLLT-229047
CELLTREAT
CELLTREAT Pipette
1000µL Pipette Tips, Bulk Packed, Non-sterile
Shipped & Sold By CELLTREAT
A Trusted Marketplace Seller
Synonym(s):
Pipette tips
Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
Marketplace Support
Need help or have questions about this product?
Contact Seller
manufacturer/tradename
CELLTREAT
General description
  • Bulk, non-sterile Pipette Tips packed in unique self-standing, resealable bags
  • CELLTREAT Scientific Products Pipette Tips deliver high performance with exceptional value. A perfected balance between raw materials and efficient manufacturing processes results in superior low retention
  • Pipette Tips have smooth, hydrophobic surfaces to ensure accurate pipetting
  • Multiple Pipette Tip sizes are available, including extended length tips (10μL and 1000μL) to allow better access to the bottom of test tubes, reagent bottles, flasks and other vessels
  • 200μL filter tip is longer than standard tips for better access to vessels and increased headspace to filter
  • Pipette Tips are universal fit and compatible with major pipettes
  • Fine tip for improved accuracy and precision
  • Molded volume reference markings offer a quick double check opportunity
Still not finding the right product?
Explore all of our products under CELLTREAT Pipette
Choose from one of the most recent versions:
Certificates of Analysis (COA)