MilliporeSigma
CLLT-229054
CELLTREAT
CELLTREAT Pipette tips
200µL Low Retention Pipette Tip Reload System, Non-sterile
Shipped & Sold By CELLTREAT
A Trusted Marketplace Seller
Synonym(s):
Low retention pipette tips
Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
Marketplace Support
Need help or have questions about this product?
Contact Seller
manufacturer/tradename
CELLTREAT
General description
  • Reload System offers clear window for easy identification of color-coded wafers
  • Support wafers have large grasping tabs for easy removal from Reload System and Rack
  • Secure locking tab keeps reload system closed and prevents tumbling tips
  • Reduce plastic waste and save time by refilling empty racks
  • CELLTREAT Scientific Products Pipette Tips deliver high performance with exceptional value. A perfected balance between raw materials and efficient manufacturing processes results in superior low retention
  • Pipette Tips have smooth, hydrophobic surfaces to ensure accurate pipetting
  • Multiple Pipette Tip sizes are available, including extended length tips (10μL and 1000μL) to allow better access to the bottom of test tubes, reagent bottles, flasks and other vessels
  • Pipette Tips are universal fit and compatible with major pipettes
  • Fine tip for improved accuracy and precision
  • Molded volume reference markings offer a quick double-check opportunity
Still not finding the right product?
Explore all of our products under CELLTREAT Pipette tips
Choose from one of the most recent versions:
Certificates of Analysis (COA)