MilliporeSigma
CLLT-229452
CELLTREAT
CELLTREAT Centrifuge tubes
TUBE & CAP, 15mL Centrifuge Tube & Cap - Bags, Non-sterile (Tubes and Caps Packed Separately)
Shipped & Sold By CELLTREAT
A Trusted Marketplace Seller
Synonym(s):
Centrifuge tube caps
Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
Marketplace Support
Need help or have questions about this product?
Contact Seller
manufacturer/tradename
CELLTREAT
General description
  • Centrifuge Tubes and Caps packed separately
  • Graduations are printed in bold, easy-to-read black ink and are accurate to +/-2%
  • Large white marking area provides excellent contrast for identification
  • Easy to write on flat surface polyethylene (HDPE) caps feature integrated leak-proof seal
  • Tubes include graduations molded into the conical bottom, ideal for pellet viewing and small volume quantification
Still not finding the right product?
Explore all of our products under CELLTREAT Centrifuge tubes
Choose from one of the most recent versions:
Certificates of Analysis (COA)