MilliporeSigma
CLLT-230299
CELLTREAT
CELLTREAT Pipettes
1-10µL Pipette, Adjustable Volume, Single Channel
Shipped & Sold By CELLTREAT
A Trusted Marketplace Seller
Synonym(s):
Pipettors
Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
Marketplace Support
Need help or have questions about this product?
Contact Seller
manufacturer/tradename
CELLTREAT
General description
  • High precision adjustable pipette with easy-to-read volume display
  • Continuously adjustable with locking ring to prevent volume changes while pipetting
  • Lightweight design and easy operation with either hand
  • Reduced operating forces; plunger is easy to press and dispense
  • Stainless steel, PTFE and PVDF components for long life
  • It is recommended to not autoclave more than 6 times
  • Fits CELLTREAT Scientific Products Universal Pipette Tips for accurate pipetting
Still not finding the right product?
Explore all of our products under CELLTREAT Pipettes
Choose from one of the most recent versions:
Certificates of Analysis (COA)