MilliporeSigma
CLLT-9025
CELLTREAT
VistaLab Pipette Tips
250µL Pipette Tips, MLA, Stacked Dense Rack, Non-sterile
Shipped & Sold By CELLTREAT
A Trusted Marketplace Seller
Synonym(s):
pipette tips
Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
Marketplace Support
Need help or have questions about this product?
Contact Seller
manufacturer/tradename
CELLTREAT
General description
A superior fit for your MLA & Ovation Pipettes. Use genuine VistaLab pipette tips to maximize the exceptional performance and benefits of Ovation and MLA pipettes. All VistaLab pipettes are calibrated with VistaLab tips to provide a perfect fit and deliver reliable performance every time.
Stacked Rack
  • Non-sterile only
  • Tips and racks are manufactured from 100% virgin polypropylene and can be autoclaved
  • 200 tips/layer
  • For Small volume (250 mL) tips - 5 layers - 1000 tips
  • For Large volume (1250 mL) tips - 3 layers - 600 tips
Still not finding the right product?
Explore all of our products under VistaLab Pipette Tips
Choose from one of the most recent versions:
Certificates of Analysis (COA)