MilliporeSigma
CLLT-9048
CELLTREAT
VistaLab Pipette Tips
5mL Pipette Tips, MLA, Graduated, Boxed, Non-sterile
Shipped & Sold By CELLTREAT
A Trusted Marketplace Seller
Synonym(s):
pipette tips
Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
Marketplace Support
Need help or have questions about this product?
Contact Seller
manufacturer/tradename
CELLTREAT
General description
A superior fit for your MLA & Ovation Pipettes. Use genuine VistaLab pipette tips to maximize the exceptional performance and benefits of Ovation and MLA pipettes. All VistaLab pipettes are calibrated with VistaLab tips to provide a perfect fit and deliver reliable performance every time.
Macro Volume
  • Offer improved accuracy over serological pipets
  • Tip length allows better access to the bottom of test tubes, reagent bottles, flasks and other vessels
  • 5mL macro tips are graduated at 0.1mL increments
  • Available sterile and non-sterile in a variety of packaging options: Box, VistaClear™ Box, VistaBulk™, and VistaTip™
  • Tips are manufactured from 100% virgin polypropylene and can be autoclaved
  • VistaClear Box Sterile, filtered are certified RNase/DNase free & Non-Pyrogenic
Still not finding the right product?
Explore all of our products under VistaLab Pipette Tips
Choose from one of the most recent versions:
Certificates of Analysis (COA)