Skip to Content
MilliporeSigma

CH4424847503

ChemBridge Corporation

methyl (4S)-4-amino-1-methyl-L-prolinate dihydrochloride

Synonym(s):

methyl (2S,4S)-4-amino-1-methyl-2-pyrrolidinecarboxylate dihydrochloride

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

CAS Number:
Molecular Weight:
231.12
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C7H14N2O2.2ClH/c1-9-4-5(8)3-6(9)7(10)11-2;;/h5-6H,3-4,8H2,1-2H3;2*1H/t5-,6-;;/m0../s1

Inchi Key

KRVAEZGAEYMPOS-USPAICOZSA-N

Storage temp.

RT

Salt Data

2HCl

Purity

95%

Country of Origin

KZ

Shipping Temperature

RT

Physical Form

solid



MSDS

https://www.hit2lead.com/sds/4036527_sds.pdf



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library