Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
Molecular Weight:
340.36
MDL number:
Inchi Code
1S/C14H16N2O6S/c1-9-14(10(2)22-15-9)23(19,20)16(8-13(17)18)11-4-6-12(21-3)7-5-11/h4-7H,8H2,1-3H3,(H,17,18)
Inchi Key
UURJADWJQZKHSL-UHFFFAOYSA-N
Physical Form
Solid
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.