Skip to Content
MilliporeSigma

CV2H2F6257BC

ChemDiv, Inc

2,9-dimethyl-1-oxo-2,9-dihydro-1H-pyrido[3,4-b]indole-4-carboxylic acid

Synonym(s):

2,9-dimethyl-1-oxo-2,9-dihydro-1H-beta-carboline-4-carboxylic acid

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Molecular Weight:
256.26
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C14H12N2O3/c1-15-7-9(14(18)19)11-8-5-3-4-6-10(8)16(2)12(11)13(15)17/h3-7H,1-2H3,(H,18,19)

Inchi Key

CBJCQWKWCLTLIS-UHFFFAOYSA-N

Physical Form

Solid





Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library