CH6H9A56B19E
Chem Impex International, Inc.
Calcium D-saccharate tetrahydrate
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Molecular Weight:
268.23
MDL number:
Inchi Code
1S/C6H10O8.Ca.H2O/c7-1(3(9)5(11)12)2(8)4(10)6(13)14;;/h1-4,7-10H,(H,11,12)(H,13,14);;1H2
InChI key
IODUFNNVCYKZRM-UHFFFAOYSA-N
storage temp.
Store at RT
Purity
98.5-102.0%
Country of Origin
US
Shipping Temperature
RT
Physical Form
White crystalline powder
Product Link
https://www.chemimpex.com/product/productinfo/calcium-d-saccharate-tetrahydrate/538?cid=0
MSDS
https://www.chemimpex.com/product/productinfo/calcium-d-saccharate-tetrahydrate/538?cid=0
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service