Inchi Code
1S/C4H10N3O5P/c1-7(4(5)6)2-3(8)12-13(9,10)11/h2H2,1H3,(H3,5,6)(H2,9,10,11)/p-1
Inchi Key
KPXOWQOQJKGYRN-UHFFFAOYSA-M
Purity
97%.
Country of Origin
US
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.