Inchi Code
1S/C25H27N9O8S2.Na/c1-3-32-8-9-33(21(39)20(32)38)24(42)27-15(12-4-6-14(35)7-5-12)18(36)26-16-19(37)34-17(23(40)41)13(10-43-22(16)34)11-44-25-28-29-30-31(25)2;/h4-7,15-16,22,35H,3,8-11H2,1-2H3,(H,26,36)(H,27,42)(H,40,41);
InChI key
NMANBWFYJSWTBZ-UHFFFAOYSA-N
storage temp.
Store at 0-8 C
Purity
870-1015
Country of Origin
US
Shipping Temperature
RT
Product Link
https://www.chemimpex.com/products/00439
MSDS
https://www.chemimpex.com/products/00439
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service