Skip to Content
MilliporeSigma

CH6H9A56B3B6

Chem Impex International, Inc.

Oleic acid

Synonym(s):

octadec-9-enoic acid

Sign In to View Organizational & Contract Pricing.

Select a Size


Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

Inchi Code

1S/C18H34O2/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18(19)20/h9-10H,2-8,11-17H2,1H3,(H,19,20)

Inchi Key

ZQPPMHVWECSIRJ-UHFFFAOYSA-N

Shipping Temperature

RT

Purity

99%

Storage temp.

Store at 0-8 C

Country of Origin

US

Physical Form

Colorless to pale yellow liquid

Boiling Point

194-195C/1.2mm


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service