Skip to Content
MilliporeSigma

CH6H9A56B8AB

Chem Impex International, Inc.

Fmoc-3-fluoro-L-phenylalanine

Synonym(s):

2-(((9H-fluoren-9-yl)methoxy)carbonylamino)-3-(3-fluorophenyl)propanoic acid

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C24H20O4N1F1
CAS Number:
Molecular Weight:
405.43
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C24H20FNO4/c25-16-7-5-6-15(12-16)13-22(23(27)28)26-24(29)30-14-21-19-10-3-1-8-17(19)18-9-2-4-11-20(18)21/h1-12,21-22H,13-14H2,(H,26,29)(H,27,28)

Inchi Key

DWSDVARCJDOADL-UHFFFAOYSA-N

Shipping Temperature

RT

Purity

98%

Storage temp.

Store at 0-8 C

Country of Origin

US

Physical Form

White to off-white powder

Melting Point

150 - 161 C



Still not finding the right product?

Explore all of our products under Fmoc-3-fluoro-L-phenylalanine




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library