Skip to Content
MilliporeSigma
All Photos(1)

Documents

CH6H9A56C680

Chem Impex International, Inc.

L-Serine benzyl ester hydrochloride

Sign Into View Organizational & Contract Pricing

CAS Number:
Molecular Weight:
231.68
MDL number:

mp

170 - 178 C

Iupac Name

benzyl 2-amino-3-hydroxypropanoate hydrochloride

Inchi Code

1S/C10H13NO3.ClH/c11-9(6-12)10(13)14-7-8-4-2-1-3-5-8;/h1-5,9,12H,6-7,11H2;1H

InChI key

MGZWCDQAKCHOBX-UHFFFAOYSA-N

storage temp.

Store at 0 - 8 C

Purity

99%

Shipping Temperature

RT

Physical Form

White to off-white powder

Product Link

https://www.chemimpex.com/product/productinfo/l-serine-benzyl-ester-hydrochloride/4351?cid=0

Melting Point

170 - 178 C

MSDS

https://www.chemimpex.com/product/productinfo/l-serine-benzyl-ester-hydrochloride/4351?cid=0


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service