Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

CH6H9A56D603

Chem Impex International, Inc.

New Coccine

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
604.48
MDL number:

Iupac Name

trisodium 7-hydroxy-8-[(E)-(4-sulfonato-1-naphthyl)diazenyl]-1,3-naphthalenedisulfonate

Inchi Code

1S/C20H14N2O10S3.3Na/c23-16-7-5-11-9-12(33(24,25)26)10-18(35(30,31)32)19(11)20(16)22-21-15-6-8-17(34(27,28)29)14-4-2-1-3-13(14)15;;;/h1-10,23H,(H,24,25,26)(H,27,28,29)(H,30,31,32);;;/q;3*+1/p-3

InChI key

SWGJCIMEBVHMTA-UHFFFAOYSA-K

storage temp.

Store at RT

Purity

84%

Country of Origin

US

Shipping Temperature

RT

Physical Form

Red to dark red to brown powder

Product Link

https://www.chemimpex.com/product/productinfo/new-coccine/13385?cid=0

MSDS

https://www.chemimpex.com/product/productinfo/new-coccine/13385?cid=0


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service