Skip to Content
MilliporeSigma

CH6H9A56E97B

Chem Impex International, Inc.

Tributylammonium pyrophosphate

Synonym(s):

tributylamine diphosphate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C12H31O7N1P2
CAS Number:
Molecular Weight:
363.33
MDL number:


Inchi Code

1S/C12H27N.H4O7P2/c1-4-7-10-13(11-8-5-2)12-9-6-3;1-8(2,3)7-9(4,5)6/h4-12H2,1-3H3;(H2,1,2,3)(H2,4,5,6)

Inchi Key

WJFREWBMJPXLBS-UHFFFAOYSA-N

Purity

99%31

Country of Origin

US





Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library