Skip to Content
MilliporeSigma

CIAH987EF3B7

ChemScene LLC (Preferred Partner)

Ethyl 3-oxo-3-phenylpropanoate

Synonym(s):

ethyl 3-oxo-3-phenylpropanoate

Sign In to View Organizational & Contract Pricing.

Select a Size


About This Item

CAS Number:
Molecular Weight:
192.21
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

bp

268.1C at 760 mmHg

mp

<0 C

Inchi Code

1S/C11H12O3/c1-2-14-11(13)8-10(12)9-6-4-3-5-7-9/h3-7H,2,8H2,1H3

Inchi Key

GKKZMYDNDDMXSE-UHFFFAOYSA-N

Storage temp.

Store at room temperature

Purity

95%

Country of Origin

CN

Physical Form

Liquid

Flash Point

140C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service