Skip to Content
MilliporeSigma

CIAH987EF89C

ChemScene LLC (Preferred Partner)

Naphthalen-1-ylhydrazine hydrochloride

Synonym(s):

1-(1-naphthyl)hydrazine hydrochloride

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C10H11N2Cl1
CAS Number:
Molecular Weight:
194.66
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C10H10N2.ClH/c11-12-10-7-3-5-8-4-1-2-6-9(8)10;/h1-7,12H,11H2;1H

Inchi Key

FYSSYOCJFZSKNW-UHFFFAOYSA-N

Purity

97%

Salt Data

Hydrochloride

Storage temp.

4C, stored under nitrogen

Country of Origin

CN

Physical Form

Solid

Melting Point

119 C



Still not finding the right product?



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library