Skip to Content
MilliporeSigma

CIAH987EFD6F

ChemScene LLC (Preferred Partner)

3,5-Dichloropyridine

Synonym(s):

3,5-dichloropyridine

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C5H3N1Cl2
CAS Number:
Molecular Weight:
147.99
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C5H3Cl2N/c6-4-1-5(7)3-8-2-4/h1-3H

Inchi Key

WPGHPGAUFIJVJF-UHFFFAOYSA-N

Purity

98%

Storage temp.

4C

Country of Origin

CN

Physical Form

Solid

Boiling Point

173.7C at 760 mmHg

Melting Point

65-67C

Density

1.39g/cm3



Still not finding the right product?

Explore all of our products under 3,5-Dichloropyridine



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library