Skip to Content
MilliporeSigma

CIAH987EFD9F

ChemScene LLC (Preferred Partner)

1-Methylpiperidin-4-amine

Synonym(s):

1-methyl-4-piperidinylamine

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C6H14N2
CAS Number:
Molecular Weight:
114.19
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C6H14N2/c1-8-4-2-6(7)3-5-8/h6H,2-5,7H2,1H3

Inchi Key

ALOCUZOKRULSAA-UHFFFAOYSA-N

Purity

98%

Storage temp.

Store at room temperature

Country of Origin

CN

Boiling Point

141C at 760 mmHg

Melting Point

203-207C

Flash Point

39.2C

Physical Form

Liquid



Still not finding the right product?

Explore all of our products under 1-Methylpiperidin-4-amine


hstatements

signalword

Danger



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library