Skip to Content
MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now

CIAH987F0B61

ChemScene LLC (Preferred Partner)

POPSO,99% with 2H2O

Synonym(s):

3,3'-(piperazine-1,4-diyl)bis(2-hydroxypropane-1-sulfonic acid)

Sign In to View Organizational & Contract Pricing.

Select a Size


About This Item

Linear Formula:
C10H22O8N2S2
CAS Number:
Molecular Weight:
362.43
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

Inchi Code

1S/C10H22N2O8S2/c13-9(7-21(15,16)17)5-11-1-2-12(4-3-11)6-10(14)8-22(18,19)20/h9-10,13-14H,1-8H2,(H,15,16,17)(H,18,19,20)

Inchi Key

LVQFQZZGTZFUNF-UHFFFAOYSA-N

Purity

99%22

Storage temp.

4C

Country of Origin

CN

Physical Form

Solid

Melting Point

320 C

Looking for similar products? Visit Product Comparison Guide


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service