Skip to Content
MilliporeSigma

CIAH987F4DD2

ChemScene LLC (Preferred Partner)

(3aS,4R,6S,6aR)-6-(2-((tert-butyldimethylsilyl)oxy)ethoxy)-2,2-dimethyltetrahydro-3aH-cyclopenta[d][1,3]dioxol-4-amine

Synonym(s):

(3aS,4R,6S,6aR)-6-(2-((tert-butyldimethylsilyl)oxy)ethoxy)-2,2-dimethyltetrahydro-4H-cyclopenta[d][1,3]dioxol-4-amine

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C16H33O4N1Si1
CAS Number:
Molecular Weight:
331.53
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C16H33NO4Si/c1-15(2,3)22(6,7)19-9-8-18-12-10-11(17)13-14(12)21-16(4,5)20-13/h11-14H,8-10,17H2,1-7H3/t11-,12+,13+,14-/m1/s1

Inchi Key

UGMXRTTVIZNRAK-ZOBORPQBSA-N

Purity

98%

Storage temp.

4C

Country of Origin

CN






Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library