Skip to Content
MilliporeSigma

CIAH98D204D9

ChemScene LLC (Preferred Partner)

2-((Carboxymethyl)amino)benzoic acid

Synonym(s):

2-[(carboxymethyl)amino]benzoic acid

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C9H9O4N1
CAS Number:
Molecular Weight:
195.17
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C9H9NO4/c11-8(12)5-10-7-4-2-1-3-6(7)9(13)14/h1-4,10H,5H2,(H,11,12)(H,13,14)

Inchi Key

PJUXPMVQAZLJEX-UHFFFAOYSA-N

Purity

98%

Storage temp.

4C

Country of Origin

CN

Physical Form

Solid

Boiling Point

484.7 C at 760 mmHg

Melting Point

218C



Still not finding the right product?




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library