Skip to Content
MilliporeSigma

CIAH991799A4

ChemScene LLC (Preferred Partner)

Dodecane-1,12-diol

Synonym(s):

1,12-dodecanediol

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C12H26O2
CAS Number:
Molecular Weight:
202.34
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C12H26O2/c13-11-9-7-5-3-1-2-4-6-8-10-12-14/h13-14H,1-12H2

Inchi Key

GHLKSLMMWAKNBM-UHFFFAOYSA-N

Purity

97%

Storage temp.

4C

Country of Origin

CN

Physical Form

Solid

Boiling Point

313.3C at 760 mmHg

Melting Point

81-84C

Flash Point

176C



Still not finding the right product?

Explore all of our products under Dodecane-1,12-diol



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library