Skip to Content
MilliporeSigma

CIAH9A904F44

ChemScene LLC (Preferred Partner)

EDTA (disodium)

Synonym(s):

sodium 2,2'-(2-(bis(carboxymethyl)amino)ethylazanediyl)diacetate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C10H14O8N2Na2
CAS Number:
Molecular Weight:
336.21
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C10H16N2O8.2Na/c13-7(14)3-11(4-8(15)16)1-2-12(5-9(17)18)6-10(19)20;;/h1-6H2,(H,13,14)(H,15,16)(H,17,18)(H,19,20);;/q;2*+1/p-2

Inchi Key

ZGTMUACCHSMWAC-UHFFFAOYSA-L

Purity

98%

Salt Data

Sodium

Storage temp.

Store at room temperature

Country of Origin

CN

Physical Form

Solid

Boiling Point

>100 C at 760 mmHg

Melting Point

248C

Density

1.01g/cm3



hstatements

signalword

Warning



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library