Skip to Content
MilliporeSigma

CIAH9A9B853B

ChemScene LLC (Preferred Partner)

(E)-β-Ionone

Synonym(s):

(3E)-4-(2,6,6-trimethyl-1-cyclohexen-1-yl)-3-buten-2-one

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C13H20O1
CAS Number:
Molecular Weight:
192.3
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C13H20O/c1-10-6-5-9-13(3,4)12(10)8-7-11(2)14/h7-8H,5-6,9H2,1-4H3/b8-7+

Inchi Key

PSQYTAPXSHCGMF-BQYQJAHWSA-N

Purity

96%

Storage temp.

Store at room temperature

Country of Origin

CN

Melting Point

-49 C

Physical Form

Liquid

Density

0.9385g/cm3



hstatements

signalword

Warning



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library