Skip to Content
MilliporeSigma

CIAH9BC06A37

ChemScene LLC (Preferred Partner)

N2-iBu-5'-O-DMT-2'-O-TBDMS-G-CE, 99%, ���0.2%(K.F.)

Synonym(s):

(2R,5R)-2-((bis(4-methoxyphenyl)(phenyl)methoxy)methyl)-4-(tert-butyldimethylsilyloxy)-5-(6-hydroxy-2-isobutyramido-9H-purin-9-yl)tetrahydrofuran-3-yl 2-cyanoethyl diisopropylphosphoramidite

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C50H68N7O9PSi/c1-32(2)45(58)54-48-53-44-41(46(59)55-48)52-31-56(44)47-43(66-68(12,13)49(7,8)9)42(65-67(63-29-17-28-51)57(33(3)4)34(5)6)40(64-47)30-62-50(35-18-15-14-16-19-35,36-20-24-38(60-10)25-21-36)37-22-26-39(61-11)27-23-37/h14-16,18-27,31-34,40,42-43,47H,17,29-30H2,1-13H3,(H2,53,54,55,58,59)/t40-,42?,43?,47-,67?/m1/s1

Inchi Key

PGJKESPHANLWME-RXAZPWLISA-N

Purity

99%0.2%..

Storage temp.

-20C

Country of Origin

CN





Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library