Skip to Content
MilliporeSigma

COMH93D672F4

Combi-Blocks, Inc.

Vinyl laurate

Synonym(s):

vinyl laurate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

CAS Number:
Molecular Weight:
226.36
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C14H26O2/c1-3-5-6-7-8-9-10-11-12-13-14(15)16-4-2/h4H,2-3,5-13H2,1H3

Inchi Key

GLVVKKSPKXTQRB-UHFFFAOYSA-N

Purity

98%

Country of Origin

CN



MSDS

https://www.combi-blocks.com/cgi-bin/find.cgi?QH-0357



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library