Skip to Content
MilliporeSigma

COMH9A888AB1

Combi-Blocks, Inc.

Azepan-2-one hydrate

Synonym(s):

2-azepanone hydrate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C6H13O2N1
CAS Number:
Molecular Weight:
131.17
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C6H11NO.H2O/c8-6-4-2-1-3-5-7-6;/h1-5H2,(H,7,8);1H2

Inchi Key

AWSFEOSAIZJXLG-UHFFFAOYSA-N

Purity

99%

Country of Origin

CN





Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library