Skip to Content
MilliporeSigma

SWC-002R

CP Lab Safety

CP Lab Safety Bench Top Solid Waste Container with Base

5 Liter, Red Lid, Unlabeled

Shipped & Sold By CP Lab Safety

A Trusted Marketplace Seller

Synonym(s):

trash can

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

Marketplace Support
Need help or have questions about this product?
Contact Seller


material

polyethylene

manufacturer/tradename

CP Lab Safety

General description

Keep your lab RCRA compliant by segregating hazardous solid waste from normal waste in our 5 liter Bench Top Solid Waste Container.

It is against the law to add chemically contaminated solid waste to the general trash in the laboratory. Our Bench Top Solid Waste Container with lid is a convenient way to discard of contaminated solid wastes right where you work. Not carrying your contaminated items across a lab to a larger waste container means preventing cross contamination. The latching lid reduces solvent emissions from the waste inside, keeping the air you breathe safer and healthier.

Our Benchtop Solid Waste Container with detachable base and lid with gasket is an ideal containment system for contaminated solid wastes, preventing possible cross contamination of products. The latching lid significantly reduces the emission of solvent-soaked solid waste back into the workspace. ideal if you routinely handle radioisotopes, beta-emitters, wet silica gel, contaminated gloves, kim-wipes, filter papers, wiping rags, pipets, pipet bulbs, and other disposables.

It has a small footprint and can easily sit in the corner of a fume hood or bench, and is made of chemically resistant HDPE with a detachable base. Shipped with a sample liner bag. Additional can liners are sold separately in packs of 100 or 250, red or yellow or biohazard bags.

Product Specifications

  • Height (Lid Closed): 11 in.
  • Height (Lid Open): 21 in.
  • Footprint (with base): 13 x 13 in.
  • Footprint (without base): 7 x 7 in.
  • Capacity (approx.) 1.5G / 5L


Still not finding the right product?




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library