Iupac Name
sulfuric acid compound with 1-pyrrolidinecarboximidamide (1:1)
Inchi Code
1S/C5H11N3.H2O4S/c6-5(7)8-3-1-2-4-8;1-5(2,3)4/h1-4H2,(H3,6,7);(H2,1,2,3,4)
InChI key
YFQLHYLNOMNMMW-UHFFFAOYSA-N
storage temp.
RT
Salt Data
O=S(=O)(O)O
Purity
95
Country of Origin
UA
Shipping Temperature
Normal
Product Link
https://www.enaminestore.com/catalog/EN300-23620
Signal Word
Warning
Hazard Statements
MSDS
https://www.enaminestore.com/catalog/EN300-23620
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service